Assay Details
Target Gene Details
Entrez Gene ID: | 30820 |
Gene Name: | potassium voltage-gated channel interacting protein 1 |
Gene Aliases: |
KCHIP1, VABP |
Location: |
Chr.5:170353055-170736632 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNIP1 | NM_001034838.2 | NP_001030010.1 | ||
| XM_017009407.1 | XP_016864896.1 | |||
| XM_017009408.1 | XP_016864897.1 | |||
| AK301775.1 | ||||
| DC420716.1 | ||||
| DQ148476.1 | AAZ77793.1 |
Target Gene Details
Entrez Gene ID: | 3779 |
Gene Name: | potassium calcium-activated channel subfamily M regulatory beta subunit 1 |
Gene Aliases: |
BKbeta1, K(VCA)beta, SLO-BETA, hbeta1, hslo-beta, k(VCA)beta-1, slo-beta-1 |
Location: |
Chr.5:170378161-170389677 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNMB1 | NM_004137.3 | 4 | 851 | NP_004128.1 |
| AF026002.1 | 3 | 408 | AAB88805.1 | |
| AK313979.1 | 4 | 829 | ||
| AY044441.1 | 4 | 474 | AAK95827.1 | |
| AY515264.1 | 4 | 512 | AAS20193.1 | |
| U25138.1 | 4 | 567 | AAB02394.1 | |
| U38907.1 | 3 | 408 | AAA81327.1 | |
| U42600.1 | 4 | 461 | AAB16827.1 | |
| U61536.1 | 4 | 513 | AAB61396.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv471054 | Chr.5:170278287 - 170532433 on Build GRCh38 | Loss |
|
| nsv462515 | Chr.5:170265550 - 170532433 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76348543] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map