Assay Details
Target Gene Details
Entrez Gene ID: | 163702 |
Gene Name: | interferon lambda receptor 1 |
Gene Aliases: |
CRF2/12, IFNLR, IL-28R1, IL28RA, LICR2 |
Location: |
Chr.1:24154156-24187275 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IFNLR1 | NM_170743.3 | 7 | 3185 | NP_734464.1 |
| NM_173064.2 | 7 | 3098 | NP_775087.1 | |
| NM_173065.2 | 6 | 3054 | NP_775088.1 | |
| XM_006710394.3 | 6 | 3249 | XP_006710457.2 | |
| XM_017000479.1 | 6 | 3214 | XP_016855968.1 | |
| AF439325.1 | 7 | 3158 | AAN63632.1 | |
| AK160364.1 | 7 | 3161 | ||
| BC140872.1 | 7 | 3234 |
More Information
Set Membership: |
|

Genomic Map