Assay Details
Target Gene Details
Entrez Gene ID: | 253982 |
Gene Name: | aspartate beta-hydroxylase domain containing 1 |
Gene Aliases: |
- |
Location: |
Chr.16:29900392-29919860 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASPHD1 | NM_181718.3 | NP_859069.2 | ||
| XM_017023107.1 | XP_016878596.1 | |||
| AF070642.1 | ||||
| BC009033.1 | AAH09033.1 | |||
| BC029136.1 | AAH29136.3 | |||
| BC040933.2 | AAH40933.3 | |||
| BC085011.1 | AAH85011.1 | |||
| BC126319.1 | ||||
| BC126321.1 | ||||
| BC144152.1 |
More Information
Set Membership: |
|

Genomic Map