Assay Details
Target Gene Details
Entrez Gene ID: | 3221 |
Gene Name: | homeobox C4 |
Gene Aliases: |
HOX3, HOX3E, cp19 |
Location: |
Chr.12:54016852-54056030 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HOXC4 | NM_014620.5 | NP_055435.2 | ||
| BQ893403.1 | ||||
| X07495.1 | CAA30376.1 |
Target Gene Details
Entrez Gene ID: | 3222 |
Gene Name: | homeobox C5 |
Gene Aliases: |
CP11, HOX3, HOX3D |
Location: |
Chr.12:54016852-54035361 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HOXC5 | NM_018953.3 | 2 | 907 | NP_061826.1 |
| NR_003084.2 | 2 | 941 | ||
| BE255323.1 | 1 | 187 | ||
| CN309383.1 | 2 | 483 | ||
| X52402.1 | 1 | 316 | CAA36647.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428282 | Chr.12:53953285 - 54140862 on Build GRCh38 | Gain |
|
| nsv826379 | Chr.12:53935085 - 54056069 on Build GRCh38 | Gain |
|
| nsv832418 | Chr.12:54016556 - 54186660 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map