Assay Details
Target Gene Details
Entrez Gene ID: | 8288 |
Gene Name: | eosinophil peroxidase |
Gene Aliases: |
EPO, EPP, EPX-PEN, EPXD |
Location: |
Chr.17:58192724-58205174 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EPX | NM_000502.5 | 3 | 392 | NP_000493.1 |
| AL597635.1 | 2 | 127 | ||
| BM147889.1 | ||||
| X14346.1 | 3 | 233 | CAA32530.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv517229 | Chr.17:58176226 - 58373163 on Build GRCh38 | Gain+Loss |
|
| nsv575797 | Chr.17:58180623 - 58212973 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map