Assay Details
Target Gene Details
Entrez Gene ID: | 1791 |
Gene Name: | DNA nucleotidylexotransferase |
Gene Aliases: |
TDT |
Location: |
Chr.10:96304328-96338564 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNTT | NM_001017520.1 | 1 | 254 | NP_001017520.1 |
| NM_004088.3 | 1 | 254 | NP_004079.3 | |
| AB046378.1 | 1 | 186 | BAB72001.1 | |
| AK223317.1 | 1 | 98 | BAD97037.1 | |
| AK313615.1 | 1 | 163 | ||
| BC012920.1 | 1 | 127 | AAH12920.1 | |
| M22968.1 | 1 | 794 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2761620 | Chr.10:96287288 - 96329992 on Build GRCh38 | Gain |
|
| nsv831949 | Chr.10:96167838 - 96385945 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map