Assay Details
Target Gene Details
Entrez Gene ID: | 101927066 |
Gene Name: | uncharacterized LOC101927066 |
Gene Aliases: |
- |
Location: |
Chr.8:96951864-97446497 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927066 | NR_125390.1 | |||
| AK091342.1 |
Target Gene Details
Entrez Gene ID: | 85453 |
Gene Name: | TSPY like 5 |
Gene Aliases: |
- |
Location: |
Chr.8:97273486-97277948 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSPYL5 | NM_033512.2 | 1 | 2732 | NP_277047.2 |
| AB051537.1 | 1 | 2671 | ||
| AK091445.1 | 1 | 2748 | ||
| AK092272.1 | 1 | 1224 | ||
| AL080059.2 | 1 | 851 | ||
| BC045630.1 | 1 | 2732 | AAH45630.1 |
More Information
Set Membership: |
|

Genomic Map