Assay Details
Target Gene Details
Entrez Gene ID: | 389400 |
Gene Name: | GDNF family receptor alpha like |
Gene Aliases: |
C6orf144, GRAL, UNQ9356, bA360D14.1 |
Location: |
Chr.6:55327469-55402493 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GFRAL | NM_207410.2 | NP_997293.2 | ||
| AY358198.1 | AAQ88565.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv603162 | Chr.6:55292014 - 55361215 on Build GRCh38 | Loss |
|
| dgv5964n100 | Chr.6:55284590 - 55378351 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map