Assay Details
Target Gene Details
Entrez Gene ID: | 2826 |
Gene Name: | C-C motif chemokine receptor 10 |
Gene Aliases: |
GPR2 |
Location: |
Chr.17:42679402-42681827 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCR10 | NM_016602.2 | 2 | 929 | NP_057686.2 |
| AF208237.1 | 2 | 925 | AAF72871.1 | |
| AF215981.1 | 2 | 929 | AAF63709.1 | |
| AY429103.1 | 2 | 925 | AAR07898.1 | |
| BC098132.1 | 2 | 929 | AAH98132.1 | |
| BC098157.1 | 2 | 929 | AAH98157.1 | |
| BC098304.1 | 2 | 929 | AAH98304.1 | |
| BC099840.1 | 2 | 929 | AAH99840.1 | |
| BM930170.1 | 1 | 283 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833450 | Chr.17:42660985 - 42825144 on Build GRCh38 | Loss |
|
| nsv953899 | Chr.17:42665283 - 42692482 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map