Assay Details
Target Gene Details
Entrez Gene ID: | 125476 |
Gene Name: | INO80 complex subunit C |
Gene Aliases: |
C18orf37, IES6, hIes6 |
Location: |
Chr.18:35468327-35497991 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INO80C | NM_001098817.1 | NP_001092287.1 | ||
| NM_194281.3 | NP_919257.2 | |||
| AK095502.1 | BAC04560.1 | |||
| AK300660.1 | ||||
| BC039404.1 | AAH39404.1 | |||
| BQ887287.1 | ||||
| BY800049.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1065113 | Chr.18:35441391 - 36065348 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map