Assay Details
Target Gene Details
Entrez Gene ID: | 64866 |
Gene Name: | CUB domain containing protein 1 |
Gene Aliases: |
CD318, SIMA135, TRASK |
Location: |
Chr.3:45082274-45146422 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDCP1 | NM_022842.4 | 9 | 5255 | NP_073753.3 |
| XM_011534024.1 | 9 | 5213 | XP_011532326.1 | |
| XM_017007070.1 | 8 | 5153 | XP_016862559.1 | |
| AF468010.1 | 9 | 5230 | AAO33397.1 | |
| AK023834.1 | 7 | 4833 | BAB14695.1 | |
| AK026329.1 | 1 | 446 | ||
| AY026461.1 | 9 | 5202 | AAK02058.1 | |
| AY167484.1 | 9 | 5212 | ||
| CD367582.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834676 | Chr.3:44928707 - 45149356 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map