Assay Details
Target Gene Details
Entrez Gene ID: | 165082 |
Gene Name: | adhesion G protein-coupled receptor F3 |
Gene Aliases: |
GPR113, PGR23 |
Location: |
Chr.2:26308173-26346817 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADGRF3 | NM_001145168.1 | 10 | 1573 | NP_001138640.1 |
| NM_001145169.1 | 9 | 1419 | NP_001138641.1 | |
| NM_001321971.1 | 9 | 1952 | NP_001308900.1 | |
| NM_001321975.1 | 10 | 1493 | NP_001308904.1 | |
| NM_153835.3 | 7 | 1559 | NP_722577.2 | |
| NR_135916.1 | 7 | 1559 | ||
| XM_011532621.2 | 9 | 1904 | XP_011530923.1 | |
| XM_011532622.2 | 9 | 1856 | XP_011530924.1 | |
| XM_011532623.2 | 9 | 1848 | XP_011530925.1 | |
| XM_017003505.1 | 9 | 1890 | XP_016858994.1 | |
| XM_017003506.1 | 9 | 2146 | XP_016858995.1 | |
| AK122995.1 | 4 | 1064 | ||
| AK293887.1 | 9 | 1419 | ||
| AY140955.1 | 10 | 1573 | AAN46669.1 | |
| AY358172.1 | 7 | 1559 | AAQ88539.1 | |
| AY358214.1 | AAQ88581.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33316 | Chr.2:25440017 - 26672800 on Build GRCh38 | Loss |
|
| nsv953758 | Chr.2:26310833 - 26320232 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map