Assay Details
Target Gene Details
Entrez Gene ID: | 722 |
Gene Name: | complement component 4 binding protein alpha |
Gene Aliases: |
C4BP, PRP |
Location: |
Chr.1:207104232-207144972 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C4BPA | NM_000715.3 | 8 | 1202 | NP_000706.1 |
| XM_005273251.1 | 8 | 1158 | XP_005273308.1 | |
| XM_005273252.4 | 8 | 1334 | XP_005273309.1 | |
| AK303793.1 | 9 | 1013 | ||
| AK303798.1 | 8 | 1152 | ||
| AK313164.1 | 8 | 1148 | ||
| BC022312.1 | 8 | 1202 | AAH22312.1 | |
| M31452.1 | 8 | 1146 | AAA36507.1 | |
| X02865.1 | 6 | 771 | CAA26617.1 |
More Information
Set Membership: |
|

Genomic Map