Assay Details
Target Gene Details
Entrez Gene ID: | 55216 |
Gene Name: | chromosome 11 open reading frame 57 |
Gene Aliases: |
- |
Location: |
Chr.11:112074244-112085150 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C11orf57 | NM_001082969.1 | NP_001076438.1 | ||
| NM_001082970.1 | NP_001076439.1 | |||
| NM_001301017.1 | NP_001287946.1 | |||
| NM_001301019.1 | NP_001287948.1 | |||
| NM_001301021.1 | NP_001287950.1 | |||
| NM_018195.3 | NP_060665.3 | |||
| NR_103469.1 | ||||
| AB096251.1 | ||||
| AK094203.1 | ||||
| AK125340.1 | BAC86137.1 | |||
| BC048133.1 | ||||
| BC072455.1 | ||||
| BG115155.1 | ||||
| BX538107.1 | CAD98020.1 | |||
| CD244552.1 | ||||
| CD706604.1 | ||||
| DA813095.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2745077 | Chr.11:111949712 - 112188677 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map