Assay Details
Target Gene Details
Entrez Gene ID: | 152579 |
Gene Name: | sec1 family domain containing 2 |
Gene Aliases: |
STXBP1L1 |
Location: |
Chr.4:52872630-53366111 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCFD2 | NM_152540.3 | NP_689753.2 | ||
| AB074276.1 | ||||
| AK024713.1 | ||||
| AK096833.1 | BAC04872.1 | |||
| AY299407.1 | AAP57259.1 | |||
| BC012458.1 | AAH12458.1 | |||
| BC021137.1 | AAH21137.2 | |||
| BC032453.1 | AAH32453.1 |
More Information
Set Membership: |
|

Genomic Map