Assay Details
Target Gene Details
Entrez Gene ID: | 130888 |
Gene Name: | F-box protein 36 |
Gene Aliases: |
Fbx36 |
Location: |
Chr.2:229922491-230013109 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FBXO36 | NM_174899.4 | 1 | 87 | NP_777559.3 |
| AK123085.1 | 1 | 276 | ||
| AK289655.1 | 1 | 72 | ||
| AK309702.1 | 1 | 65 | ||
| BC033935.1 | 1 | 80 | AAH33935.1 | |
| BF674223.1 | 1 | 88 |
Target Gene Details
Entrez Gene ID: | 9320 |
Gene Name: | thyroid hormone receptor interactor 12 |
Gene Aliases: |
TRIP-12, ULF |
Location: |
Chr.2:229763837-229923234 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRIP12 | XM_011512181.2 | XP_011510483.1 | ||
| XM_011512183.2 | XP_011510485.1 | |||
| XM_017005274.1 | XP_016860763.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv164e55 | Chr.2:229802854 - 230128418 on Build GRCh38 | Gain |
|
| esv3594584 | Chr.2:229856243 - 230039898 on Build GRCh38 | Loss |
|
| esv3575337 | Chr.2:229849253 - 230039159 on Build GRCh38 | Gain |
|
| dgv7330n54 | Chr.2:229859061 - 230036931 on Build GRCh38 | Gain |
|
| esv2751841 | Chr.2:229710778 - 229964878 on Build GRCh38 | Gain |
|
| nsv584650 | Chr.2:229820254 - 230078735 on Build GRCh38 | Gain |
|
| nsv1008431 | Chr.2:229893749 - 230085172 on Build GRCh38 | Gain |
|
| dgv4179n100 | Chr.2:229846823 - 230040510 on Build GRCh38 | Gain |
|
| nsv460118 | Chr.2:229866681 - 230035049 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map