Assay Details
Target Gene Details
Entrez Gene ID: | 92014 |
Gene Name: | solute carrier family 25 member 51 |
Gene Aliases: |
CG7943, MCART1 |
Location: |
Chr.9:37877575-37904353 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC25A51 | NM_033412.3 | NP_219480.1 | ||
| NR_024872.2 | ||||
| NR_024873.2 | ||||
| AK098185.1 | ||||
| BP363489.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469814 | Chr.9:37777286 - 37950154 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map