Assay Details
Target Gene Details
Entrez Gene ID: | 79849 |
Gene Name: | PDZ domain containing 3 |
Gene Aliases: |
IKEPP, NHERF4, PDZK2 |
Location: |
Chr.11:119185457-119190223 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDZD3 | NM_001168468.1 | NP_001161940.1 | ||
| NM_024791.3 | NP_079067.3 | |||
| NR_033122.1 | ||||
| XM_011543000.2 | XP_011541302.1 | |||
| XM_011543001.2 | XP_011541303.1 | |||
| XM_011543002.2 | XP_011541304.1 | |||
| XM_011543005.2 | XP_011541307.1 | |||
| XM_017018334.1 | XP_016873823.1 | |||
| XM_017018335.1 | XP_016873824.1 | |||
| XM_017018336.1 | XP_016873825.1 | |||
| XM_017018337.1 | XP_016873826.1 | |||
| AB094096.1 | BAC76050.1 | |||
| AK026409.1 | BAB15474.1 | |||
| AK091966.1 | BAC03780.1 | |||
| AK298333.1 | ||||
| AK315403.1 | ||||
| AY047359.1 | AAL10686.1 | |||
| BC029042.1 | AAH29042.1 | |||
| BX648391.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv556467 | Chr.11:119154221 - 119193848 on Build GRCh38 | Loss |
|
| nsv556468 | Chr.11:119185525 - 119195050 on Build GRCh38 | Loss |
|
| nsv468879 | Chr.11:119106009 - 119188695 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map