Assay Details
Target Gene Details
Entrez Gene ID: | 10307 |
Gene Name: | amyloid beta precursor protein binding family B member 3 |
Gene Aliases: |
FE65L2, SRA |
Location: |
Chr.5:140558268-140564604 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APBB3 | NM_006051.3 | 13 | 1818 | NP_006042.3 |
| NM_133172.2 | 12 | 1812 | NP_573418.2 | |
| NM_133173.2 | 13 | 1797 | NP_573419.2 | |
| NM_133174.2 | 12 | 1791 | NP_573420.2 | |
| AB018247.1 | 13 | 1489 | BAA35188.1 | |
| AB049618.1 | 13 | 1681 | BAB40806.1 | |
| AB061224.1 | BAB71767.1 | |||
| AB094990.1 | BAC22578.1 | |||
| AB209839.1 | 13 | 1525 | BAD93076.1 | |
| AF224708.1 | 12 | 1473 | AAF65172.1 | |
| AF224709.1 | 13 | 1458 | AAF65173.1 | |
| AF224710.1 | 12 | 1452 | AAF65174.1 | |
| AF224711.1 | 13 | 1479 | AAF65175.1 | |
| AK074885.1 | 8 | 1724 | ||
| AK075280.1 | 13 | 1760 | ||
| AK125244.1 | 10 | 2450 | ||
| AK304824.1 | 9 | 1818 | ||
| BC013158.1 | 13 | 1797 | AAH13158.1 | |
| BC125187.1 | 13 | 1789 | ||
| BF448559.1 | 1 | 350 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv462462 | Chr.5:140556654 - 140626201 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs145698446] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map