Assay Details
Target Gene Details
Entrez Gene ID: | 2516 |
Gene Name: | nuclear receptor subfamily 5 group A member 1 |
Gene Aliases: |
AD4BP, ELP, FTZ1, FTZF1, POF7, SF-1, SF1, SPGF8, SRXY3, hSF-1 |
Location: |
Chr.9:124481234-124507420 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NR5A1 | NM_004959.4 | 7 | 1972 | NP_004950.2 |
| XM_005251871.3 | 7 | 2000 | XP_005251928.1 | |
| XM_011518455.2 | 7 | 1850 | XP_011516757.1 | |
| AK090858.1 | 2 | 1418 | ||
| BC032501.1 | 7 | 1972 | AAH32501.1 | |
| U76388.1 | 6 | 1800 | AAB53105.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526209 | Chr.9:124470928 - 124531906 on Build GRCh38 | Gain |
|
| esv3621722 | Chr.9:124378965 - 124497390 on Build GRCh38 | Gain |
|
| nsv950942 | Chr.9:124466322 - 124513521 on Build GRCh38 | Deletion |
|
| nsv517372 | Chr.9:124467839 - 124512572 on Build GRCh38 | Loss |
|
| nsv615291 | Chr.9:124464413 - 124557925 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map