Assay Details
Target Gene Details
Entrez Gene ID: | 112 |
Gene Name: | adenylate cyclase 6 |
Gene Aliases: |
AC6, LCCS8 |
Location: |
Chr.12:48766191-48789096 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADCY6 | NM_015270.4 | 22 | 4311 | NP_056085.1 |
| XM_006719210.3 | 22 | 5269 | XP_006719273.1 | |
| XM_017018743.1 | 18 | 4473 | XP_016874232.1 | |
| AB007882.2 | 21 | 4093 | ||
| AF250226.1 | 21 | 4810 | AAF82478.1 | |
| BX537664.1 | 7 | 2970 |
More Information
Set Membership: |
|

Genomic Map