Assay Details
Target Gene Details
Entrez Gene ID: | 200373 |
Gene Name: | cilia and flagella associated protein 221 |
Gene Aliases: |
FAP221, PCDP1 |
Location: |
Chr.2:119544392-119688610 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CFAP221 |
Target Gene Details
Entrez Gene ID: | 80775 |
Gene Name: | transmembrane protein 177 |
Gene Aliases: |
- |
Location: |
Chr.2:119679167-119682118 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM177 | NM_001105198.1 | 2 | 857 | NP_001098668.1 |
| NM_001105199.1 | 2 | 465 | NP_001098669.1 | |
| NM_030577.2 | 2 | 516 | NP_085054.2 | |
| AK096890.1 | 2 | 462 | ||
| AK130088.1 | 2 | 465 | ||
| AK299078.1 | 2 | 462 | ||
| AK308503.1 | ||||
| AK311734.1 | ||||
| BC004404.1 | 2 | 504 | AAH04404.1 | |
| BG280893.1 | 2 | 452 | ||
| BG776153.1 | 2 | 487 |

Genomic Map