Assay Details
Target Gene Details
Entrez Gene ID: | 8273 |
Gene Name: | solute carrier family 10 member 3 |
Gene Aliases: |
DXS253E, P3 |
Location: |
Chr.X:154487306-154490690 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC10A3 | NM_001142391.2 | NP_001135863.1 | ||
| NM_001142392.2 | NP_001135864.1 | |||
| NM_019848.4 | NP_062822.1 | |||
| XM_005277913.4 | XP_005277970.1 | |||
| XM_006724847.3 | XP_006724910.1 | |||
| XM_006724848.3 | XP_006724911.1 | |||
| XM_011531200.2 | XP_011529502.2 | |||
| XM_011531201.2 | XP_011529503.1 | |||
| BC004966.1 | AAH04966.1 | |||
| BM981524.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33784 | Chr.X:153962665 - 155699588 on Build GRCh38 | Gain+Loss |
|
| nsv519042 | Chr.X:154082978 - 154601444 on Build GRCh38 | Gain |
|
| esv33199 | Chr.X:153065734 - 154591978 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map