Assay Details
Target Gene Details
Entrez Gene ID: | 92736 |
Gene Name: | otopetrin 2 |
Gene Aliases: |
- |
Location: |
Chr.17:74922963-74933913 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OTOP2 | NM_178160.2 | NP_835454.1 | ||
| XM_011525479.1 | XP_011523781.1 | |||
| AK000063.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833540 | Chr.17:74838971 - 75019365 on Build GRCh38 | Loss |
|
| nsv518292 | Chr.17:74810882 - 74982818 on Build GRCh38 | Loss |
|
| nsv952367 | Chr.17:74751962 - 74961105 on Build GRCh38 | Deletion |
|
| nsv457909 | Chr.17:74889500 - 74982818 on Build GRCh38 | Gain |
|
| nsv1056788 | Chr.17:74922211 - 75028689 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map