Assay Details
Target Gene Details
Entrez Gene ID: | 9533 |
Gene Name: | RNA polymerase I subunit C |
Gene Aliases: |
AC40, HLD11, RPA39, RPA40, RPA5, RPAC1, RPC40, TCS3 |
Location: |
Chr.6:43517024-43562404 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLR1C | NM_001318876.1 | 7 | 800 | NP_001305805.1 |
| NM_203290.3 | 7 | 800 | NP_976035.1 | |
| XM_011515000.2 | 7 | 741 | XP_011513302.1 | |
| AF008442.1 | 7 | 726 | AAC14354.1 | |
| AF047441.1 | 7 | 736 | AAC39892.1 | |
| AK293561.1 | ||||
| BC008118.1 | 7 | 691 | AAH08118.1 | |
| BC008863.2 | 7 | 726 | AAH08863.1 | |
| BQ575535.1 | 3 | 552 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv603000 | Chr.6:43397622 - 43524840 on Build GRCh38 | Loss |
|
| nsv818414 | Chr.6:43507736 - 43528924 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map