Assay Details
Target Gene Details
Entrez Gene ID: | 113026 |
Gene Name: | phospholipase C delta 3 |
Gene Aliases: |
PLC-delta-3 |
Location: |
Chr.17:45111640-45132533 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLCD3 | NM_133373.4 | NP_588614.1 | ||
| AB075844.1 | BAB85550.1 | |||
| AK074240.1 | BAB85029.1 | |||
| AL834392.1 | CAD39054.2 | |||
| BC010668.1 | AAH10668.2 | |||
| BC072384.1 | AAH72384.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953903 | Chr.17:45081533 - 45148633 on Build GRCh38 | Deletion |
|
| nsv527788 | Chr.17:44899824 - 45226241 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map