Assay Details
Target Gene Details
Entrez Gene ID: | 8906 |
Gene Name: | adaptor related protein complex 1 gamma 2 subunit |
Gene Aliases: |
G2AD |
Location: |
Chr.14:23559563-23569056 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Target Gene Details
Entrez Gene ID: | 102724814 |
Gene Name: | uncharacterized LOC102724814 |
Gene Aliases: |
- |
Location: |
Chr.14:23561097-23568076 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC102724814 | NR_110555.1 | 3 | 1092 | |
| AK092855.1 | 3 | 1092 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv564044 | Chr.14:23539341 - 23572415 on Build GRCh38 | Loss |
|
| nsv952817 | Chr.14:23520192 - 23582591 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map