Assay Details
Target Gene Details
Entrez Gene ID: | 79633 |
Gene Name: | FAT atypical cadherin 4 |
Gene Aliases: |
CDHF14, CDHR11, FAT-J, FATJ, HKLLS2, NBLA00548, VMLDS2 |
Location: |
Chr.4:125314913-125492932 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAT4 | NM_001291285.1 | 1 | 164 | NP_001278214.1 |
| NM_001291303.1 | 1 | 164 | NP_001278232.1 | |
| NM_024582.4 | 1 | 164 | NP_078858.4 | |
| XM_011532236.2 | 2 | 1241 | XP_011530538.1 | |
| XM_011532237.2 | XP_011530539.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830051 | Chr.4:125313740 - 125482271 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map