Assay Details
Target Gene Details
Entrez Gene ID: | 51280 |
Gene Name: | golgi membrane protein 1 |
Gene Aliases: |
C9orf155, GOLPH2, GP73, HEL46, PSEC0257, bA379P1.3 |
Location: |
Chr.9:86026141-86100201 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GOLM1 | NM_016548.3 | 10 | 1878 | NP_057632.2 |
| NM_177937.2 | 10 | 1870 | NP_808800.1 | |
| AF236056.1 | 10 | 1819 | AAF44663.1 | |
| AJ420432.1 | 1 | 101 | ||
| AK027717.1 | 10 | 1841 | ||
| AK075542.1 | 10 | 1812 | BAC11685.1 | |
| AK075557.1 | 10 | 1782 | ||
| AK092943.1 | 11 | 1758 | ||
| BC001740.1 | 10 | 1793 | AAH01740.1 | |
| EU794616.1 | 10 | 1837 |
More Information
Set Membership: |
|

Genomic Map