Assay Details
Target Gene Details
Entrez Gene ID: | 91181 |
Gene Name: | nucleoporin 210 like |
Gene Aliases: |
- |
Location: |
Chr.1:153992582-154155116 on Build GRCh38 |
Assay Gene Location: | Within Exon 40 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUP210L | NM_001159484.1 | 38 | 5344 | NP_001152956.1 |
| NM_207308.2 | 40 | 5800 | NP_997191.2 | |
| XM_011510122.1 | 39 | 5631 | XP_011508424.1 | |
| XM_011510123.1 | 39 | 5624 | XP_011508425.1 | |
| XM_017002788.1 | 39 | 5678 | XP_016858277.1 | |
| AK125924.1 | 36 | 5062 | BAC86345.1 | |
| AK128108.1 | 30 | 4379 | BAC87279.1 | |
| AL133633.1 | 16 | 2118 | CAB63757.1 | |
| AL833964.1 | 24 | 3307 | CAD38812.1 | |
| BX648132.1 | 28 | 3988 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv946401 | Chr.1:153991361 - 153992953 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map