Assay Details
Target Gene Details
Entrez Gene ID: | 342667 |
Gene Name: | SH3 and cysteine rich domain 2 |
Gene Aliases: |
24b2, 24b2/STAC2 |
Location: |
Chr.17:39210536-39225787 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STAC2 | NM_198993.3 | NP_945344.1 | ||
| XM_017024580.1 | XP_016880069.1 | |||
| XM_017024581.1 | XP_016880070.1 | |||
| XM_017024582.1 | XP_016880071.1 | |||
| XM_017024583.1 | XP_016880072.1 | |||
| AJ608762.1 | CAE75539.1 | |||
| AK131500.1 | BAD18644.1 | |||
| AM903073.1 | ||||
| BC109231.2 | AAI09232.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833441 | Chr.17:39164554 - 39320543 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map