Assay Details
Target Gene Details
Entrez Gene ID: | 5296 |
Gene Name: | phosphoinositide-3-kinase regulatory subunit 2 |
Gene Aliases: |
MPPH, MPPH1, P85B, p85, p85-BETA |
Location: |
Chr.19:18153178-18170533 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIK3R2 | NM_005027.3 | 2 | 721 | NP_005018.1 |
| NR_073517.1 | 2 | 721 | ||
| BC032647.1 | 1 | 449 | ||
| BC070082.1 | 2 | 693 | AAH70082.1 | |
| BC090249.1 | 2 | 653 | AAH90249.1 | |
| X80907.1 | 1 | 421 | CAA56868.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2429 | Chr.19:18123369 - 18168571 on Build GRCh38 | Deletion |
|
| nsv1160592 | Chr.19:18100050 - 18194768 on Build GRCh38 | Deletion |
|
| nsv952420 | Chr.19:18117791 - 18476990 on Build GRCh38 | Deletion |
|
| nsv833775 | Chr.19:18102951 - 18279264 on Build GRCh38 | Loss |
|
| nsv833773 | Chr.19:18042161 - 18192178 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map