Assay Details
Target Gene Details
Entrez Gene ID: | 7781 |
Gene Name: | solute carrier family 30 member 3 |
Gene Aliases: |
ZNT3 |
Location: |
Chr.2:27254572-27275863 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC30A3 | NM_001318949.1 | NP_001305878.1 | ||
| NM_001318950.1 | NP_001305879.1 | |||
| NM_001318951.1 | NP_001305880.1 | |||
| NM_003459.4 | NP_003450.2 | |||
| XM_005264547.1 | XP_005264604.1 | |||
| XM_005264548.3 | XP_005264605.1 | |||
| XM_006712100.2 | XP_006712163.1 | |||
| XM_011533102.2 | XP_011531404.1 | |||
| XM_011533103.2 | XP_011531405.1 | |||
| XM_017004873.1 | XP_016860362.1 | |||
| XM_017004874.1 | XP_016860363.1 | |||
| XM_017004875.1 | XP_016860364.1 | |||
| AK127096.1 | 3 | 1861 | ||
| AK127300.1 | ||||
| AK302174.1 | ||||
| AK313600.1 | ||||
| BC028358.2 | AAH28358.1 | |||
| U76010.1 | AAB39732.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833603 | Chr.2:27059106 - 27263631 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map