Assay Details
Target Gene Details
Entrez Gene ID: | 6439 |
Gene Name: | surfactant protein B |
Gene Aliases: |
PSP-B, SFTB3, SFTP3, SMDP1, SP-B |
Location: |
Chr.2:85657317-85668741 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SFTPB | NM_000542.3 | 9 | 1053 | NP_000533.3 |
| NM_198843.2 | 9 | 1053 | NP_942140.2 | |
| XM_005264487.2 | 8 | 968 | XP_005264544.1 | |
| XM_005264488.3 | 8 | 921 | XP_005264545.2 | |
| XM_005264490.4 | 8 | 933 | XP_005264547.2 | |
| XM_017004702.1 | 8 | 968 | XP_016860191.1 | |
| XM_017004703.1 | 8 | 921 | XP_016860192.1 | |
| AK303817.1 | 7 | 841 | ||
| AK314947.1 | 8 | 933 | ||
| AL548140.3 | ||||
| BC032785.1 | 8 | 920 | AAH32785.1 | |
| DQ317589.1 | 8 | 894 | ||
| J02761.1 | 8 | 930 | AAA60212.1 | |
| M16764.1 | 5 | 632 | AAA88099.1 | |
| M19097.1 | 4 | 503 | AAA36628.1 |
More Information
Set Membership: |
|

Genomic Map