Assay Details
Target Gene Details
Entrez Gene ID: | 167826 |
Gene Name: | oligodendrocyte transcription factor 3 |
Gene Aliases: |
Bhlhb7, bHLHe20 |
Location: |
Chr.6:137492199-137494394 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OLIG3 | NM_175747.2 | 1 | 1045 | NP_786923.1 |
| AK096362.1 | 1 | 1045 | BAC04768.1 | |
| BC051352.1 | 1 | 1035 | AAH51352.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5491 | Chr.6:137473055 - 137505021 on Build GRCh38 | Insertion |
|
| nsv470863 | Chr.6:137080640 - 138014159 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map