Assay Details
Target Gene Details
Entrez Gene ID: | 54438 |
Gene Name: | glucose-fructose oxidoreductase domain containing 1 |
Gene Aliases: |
ADG-90, C6orf114 |
Location: |
Chr.6:13357830-13487637 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GFOD1 | NM_001242628.1 | NP_001229557.1 | ||
| NM_001242629.1 | NP_001229558.1 | |||
| NM_018988.3 | NP_061861.1 | |||
| NR_038459.1 | ||||
| AF264036.1 | AAK58533.1 | |||
| AK000337.1 | BAA91093.1 | |||
| AK055131.1 | ||||
| BC066553.1 | AAH66553.1 | |||
| BC119005.1 | ||||
| BC124557.1 | ||||
| BC148295.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470802 | Chr.6:13441192 - 13519139 on Build GRCh38 | Gain |
|
| esv3608126 | Chr.6:13460936 - 13471362 on Build GRCh38 | Loss |
|
| nsv600994 | Chr.6:13426307 - 13519202 on Build GRCh38 | Gain |
|
| nsv1033620 | Chr.6:13419498 - 13571556 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map