Assay Details
Target Gene Details
Entrez Gene ID: | 10535 |
Gene Name: | ribonuclease H2 subunit A |
Gene Aliases: |
AGS4, JUNB, RNASEHI, RNHIA, RNHL |
Location: |
Chr.19:12802054-12813648 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNASEH2A | NM_006397.2 | 8 | 1027 | NP_006388.2 |
| XM_006722619.2 | 9 | 1212 | XP_006722682.1 | |
| AY363912.1 | 8 | 1016 | AAQ64005.1 | |
| BC011748.2 | AAH11748.1 | |||
| BM664190.1 | 1 | 130 | ||
| CB240995.1 | 1 | 130 | ||
| Z97029.1 | CAB09725.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953977 | Chr.19:12666887 - 13095486 on Build GRCh38 | Deletion |
|
| nsv1160586 | Chr.19:12597480 - 12823909 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map