Assay Details
Target Gene Details
Entrez Gene ID: | 8237 |
Gene Name: | ubiquitin specific peptidase 11 |
Gene Aliases: |
UHX1 |
Location: |
Chr.X:47232915-47248328 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP11 | NM_004651.3 | 1 | 54 | NP_004642.2 |
| AK055588.1 | ||||
| AK098394.1 | ||||
| BC140849.1 | 1 | 103 |
More Information
Set Membership: |
|

Genomic Map