Assay Details
Target Gene Details
Entrez Gene ID: | 55015 |
Gene Name: | pre-mRNA processing factor 39 |
Gene Aliases: |
- |
Location: |
Chr.14:45084099-45115601 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 10 - Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRPF39 | NM_017922.3 | NP_060392.3 | ||
| AK000673.1 | BAA91318.1 | |||
| AK001990.1 | BAA92024.1 | |||
| AK307530.1 | ||||
| BC020655.1 | ||||
| BC051886.1 | AAH51886.1 | |||
| BC125126.1 | ||||
| BC125127.1 |
Target Gene Details
Entrez Gene ID: | 100113389 |
Gene Name: | small nucleolar RNA, C/D box 127 |
Gene Aliases: |
- |
Location: |
Chr.14:45110875-45110975 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNORD127 | NR_003691.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832785 | Chr.14:45051467 - 45241538 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map