Assay Details
Target Gene Details
Entrez Gene ID: | 65123 |
Gene Name: | integrator complex subunit 3 |
Gene Aliases: |
C1orf193, C1orf60, INT3, SOSS-A, SOSSA |
Location: |
Chr.1:153728050-153774808 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INTS3 | NM_001324475.1 | 11 | 1231 | NP_001311404.1 |
| NM_023015.4 | 10 | 1572 | NP_075391.3 | |
| XM_005245459.2 | 10 | 1589 | XP_005245516.1 | |
| XM_005245461.2 | 3 | 172 | XP_005245518.1 | |
| XM_006711490.3 | 11 | 1042 | XP_006711553.1 | |
| XM_011509906.2 | 10 | 954 | XP_011508208.1 | |
| AK074979.1 | 3 | 270 | BAC11329.1 | |
| AK290025.1 | 11 | 1225 | ||
| AK298746.1 | ||||
| AK304874.1 | 4 | 661 | ||
| AL080172.1 | 4 | 381 | ||
| AL832133.1 | 9 | 2607 | CAH10398.1 | |
| BC105092.1 | 10 | 1035 | AAI05093.1 | |
| BC105094.1 | 10 | 1035 | AAI05095.1 | |
| BC116458.1 | 10 | 1567 | ||
| BX640786.1 | 10 | 1558 | CAE45876.1 | |
| BX640950.1 | 10 | 1573 | CAE45974.1 | |
| CR627233.1 | 8 | 3105 | CAH10366.1 | |
| CR749212.1 | 8 | 3099 | CAH18069.1 | |
| CR749376.1 | 9 | 3224 | CAH18229.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv547941 | Chr.1:153736430 - 153828622 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map