Assay Details
Target Gene Details
Entrez Gene ID: | 2766 |
Gene Name: | guanosine monophosphate reductase |
Gene Aliases: |
GMPR 1, GMPR1 |
Location: |
Chr.6:16238580-16295549 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GMPR | NM_006877.3 | NP_006868.3 | ||
| XM_011514508.2 | XP_011512810.1 | |||
| BC008281.1 | AAH08281.1 | |||
| M24470.1 | AAA52498.1 |
More Information
Set Membership: |
|

Genomic Map