Assay Details
Target Gene Details
Entrez Gene ID: | 10592 |
Gene Name: | structural maintenance of chromosomes 2 |
Gene Aliases: |
CAP-E, CAPE, SMC-2, SMC2L1 |
Location: |
Chr.9:104093808-104141419 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMC2 | NM_001042551.1 | NP_001036016.1 | ||
| NM_001265602.1 | NP_001252531.1 | |||
| NM_006444.2 | NP_006435.2 | |||
| XM_006716933.3 | XP_006716996.1 | |||
| XM_011518149.2 | XP_011516451.1 | |||
| XM_011518150.2 | XP_011516452.1 | |||
| XM_011518151.2 | XP_011516453.1 | |||
| XM_011518152.2 | XP_011516454.1 | |||
| XM_011518153.1 | XP_011516455.1 | |||
| XM_017014207.1 | XP_016869696.1 | |||
| XM_017014210.1 | XP_016869699.1 | |||
| XM_017014211.1 | XP_016869700.1 | |||
| AF092563.1 | AAC72360.1 | |||
| AK292599.1 | ||||
| AK307752.1 | ||||
| AL833191.1 | CAI46187.1 | |||
| BC017845.1 | ||||
| BC055081.1 | AAH55081.1 | |||
| BC061906.1 | AAH61906.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv615049 | Chr.9:103527145 - 104134528 on Build GRCh38 | Gain |
|
| nsv831676 | Chr.9:103901194 - 104112176 on Build GRCh38 | Gain |
|
| nsv615064 | Chr.9:103811066 - 104367820 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map