Assay Details
Target Gene Details
Entrez Gene ID: | 10017 |
Gene Name: | BCL2 like 10 |
Gene Aliases: |
BCL-B, Boo, Diva, bcl2-L-10 |
Location: |
Chr.15:52109263-52112775 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BCL2L10 | NM_001306168.1 | 2 | 940 | NP_001293097.1 |
| NM_020396.3 | 2 | 870 | NP_065129.1 | |
| AF326964.1 | AAK48715.1 | |||
| BC104442.1 | AAI04443.1 | |||
| BC104443.1 | AAI04444.1 | |||
| BX119239.1 | 1 | 386 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv471244 | Chr.15:52013266 - 52219268 on Build GRCh38 | Gain |
|
| nsv833005 | Chr.15:52097672 - 52249133 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map