Assay Details
Target Gene Details
Entrez Gene ID: | 51335 |
Gene Name: | neugrin, neurite outgrowth associated |
Gene Aliases: |
DSC92 |
Location: |
Chr.15:90265663-90272211 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NGRN | NM_001033088.1 | NP_001028260.2 | ||
| NR_028052.1 | ||||
| AB029315.1 | BAB21533.1 | |||
| AF225423.1 | AAG09725.1 | |||
| AF242770.1 | AAF65447.1 | |||
| AK312641.1 | ||||
| AL360142.1 | CAB96088.1 | |||
| AL834503.1 | CAD39160.1 | |||
| AY049780.1 | AAL15437.1 | |||
| BC001682.2 | AAH01682.1 | |||
| BC007222.1 | AAH07222.1 | |||
| BC009389.2 | AAH09389.1 | |||
| BC017192.2 | AAH17192.1 | |||
| BC096824.1 | AAH96824.1 | |||
| BG716404.1 | ||||
| CD242640.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833087 | Chr.15:90208675 - 90425725 on Build GRCh38 | Loss |
|
| nsv1052652 | Chr.15:90101186 - 90298961 on Build GRCh38 | Gain |
|
| esv2751541 | Chr.15:90245652 - 90457213 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map