Assay Details
Target Gene Details
Entrez Gene ID: | 3709 |
Gene Name: | inositol 1,4,5-trisphosphate receptor type 2 |
Gene Aliases: |
ANHD, CFAP48, INSP3R2, IP3R2 |
Location: |
Chr.12:26335337-26833198 on Build GRCh38 |
Assay Gene Location: | Within Exon 61 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITPR2 | NM_002223.3 | 57 | 9138 | NP_002214.2 |
| XM_017019266.1 | 58 | 9197 | XP_016874755.1 | |
| XM_017019267.1 | 58 | 8721 | XP_016874756.1 | |
| XM_017019268.1 | 57 | 8598 | XP_016874757.1 | |
| D26350.1 | 57 | 9139 | BAA05384.1 |
More Information
Set Membership: |
|

Genomic Map