Assay Details
Target Gene Details
Entrez Gene ID: | 5164 |
Gene Name: | pyruvate dehydrogenase kinase 2 |
Gene Aliases: |
PDHK2, PDKII |
Location: |
Chr.17:50094737-50111369 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDK2 | NM_001199898.1 | NP_001186827.1 | ||
| NM_001199900.1 | NP_001186829.1 | |||
| NM_002611.4 | NP_002602.2 | |||
| AK055119.1 | ||||
| AK290522.1 | ||||
| AK293600.1 | ||||
| AK297089.1 | ||||
| BC005811.2 | AAH05811.1 | |||
| BC040478.1 | AAH40478.1 | |||
| BC063137.1 | AAH63137.1 | |||
| L42451.1 | AAC42010.1 |
More Information
Set Membership: |
|

Genomic Map