Assay Details
Target Gene Details
Entrez Gene ID: | 100128568 |
Gene Name: | uncharacterized LOC100128568 |
Gene Aliases: |
- |
Location: |
Chr.19:5978403-6020363 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100128568 | NR_046376.1 |
Target Gene Details
Entrez Gene ID: | 5990 |
Gene Name: | regulatory factor X2 |
Gene Aliases: |
- |
Location: |
Chr.19:5993164-6110653 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RFX2 | NM_000635.3 | 18 | 2614 | NP_000626.2 |
| NM_134433.2 | 17 | 2539 | NP_602309.1 | |
| XM_011528171.2 | 19 | 2580 | XP_011526473.1 | |
| XM_011528172.1 | 18 | 2441 | XP_011526474.1 | |
| XM_017027107.1 | 17 | 2464 | XP_016882596.1 | |
| XM_017027108.1 | 16 | 2389 | XP_016882597.1 | |
| AK024288.1 | 15 | 2153 | ||
| AK093977.1 | 10 | 3169 | ||
| AK291196.1 | 17 | 2418 | ||
| AK302661.1 | 12 | 1602 | ||
| AK303321.1 | ||||
| BC028579.1 | 18 | 2526 | AAH28579.1 | |
| BC071571.1 | 18 | 2495 | AAH71571.1 | |
| BX648130.1 | 18 | 2449 | ||
| X76091.1 | CAA53705.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3583213 | Chr.19:5989712 - 6000302 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map