Assay Details
Target Gene Details
Entrez Gene ID: | 23178 |
Gene Name: | PAS domain containing serine/threonine kinase |
Gene Aliases: |
PASKIN, STK37 |
Location: |
Chr.2:241106099-241150347 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PASK | NM_001252119.1 | 15 | 3535 | NP_001239048.1 |
| NM_001252120.1 | 16 | 3667 | NP_001239049.1 | |
| NM_001252122.1 | 15 | 3409 | NP_001239051.1 | |
| NM_015148.3 | 15 | 3514 | NP_055963.2 | |
| XM_005246991.1 | 12 | 2849 | XP_005247048.1 | |
| XM_011510826.1 | 15 | 3490 | XP_011509128.1 | |
| XM_011510828.1 | 16 | 3591 | XP_011509130.1 | |
| XM_011510829.1 | 15 | 3532 | XP_011509131.1 | |
| XM_011510830.1 | 15 | 3533 | XP_011509132.1 | |
| XM_011510831.2 | 13 | 3697 | XP_011509133.1 | |
| XM_011510832.2 | 13 | 3676 | XP_011509134.1 | |
| XM_011510833.1 | 12 | 2870 | XP_011509135.1 | |
| XM_011510834.1 | 8 | 2593 | XP_011509136.1 | |
| XM_011510835.1 | 8 | 2183 | XP_011509137.1 | |
| XM_017003634.1 | 15 | 3758 | XP_016859123.1 | |
| XM_017003635.1 | 16 | 3570 | XP_016859124.1 | |
| XM_017003636.1 | 15 | 3526 | XP_016859125.1 | |
| AF387103.1 | 15 | 3454 | AAK69752.1 | |
| AK296945.1 | 12 | 3019 | ||
| AK302417.1 | 15 | 3365 | ||
| AK302561.1 | 12 | 2849 | ||
| BC043495.1 | 9 | 2271 | AAH43495.1 | |
| BC063585.1 | 15 | 3457 | AAH63585.1 | |
| CR749231.1 | 16 | 3668 | CAH18087.1 | |
| D50925.2 | 15 | 3466 | ||
| U79240.1 | 6 | 788 | AAB50198.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575345 | Chr.2:241086207 - 241153068 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map