Assay Details
Target Gene Details
Entrez Gene ID: | 152189 |
Gene Name: | CKLF like MARVEL transmembrane domain containing 8 |
Gene Aliases: |
CKLFSF8, CKLFSF8-V2 |
Location: |
Chr.3:32238679-32370325 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CMTM8 | NM_001320308.1 | 1 | 358 | NP_001307237.1 |
| NM_178868.4 | 1 | 358 | NP_849199.2 | |
| XM_011533416.2 | 1 | 89 | XP_011531718.1 | |
| AF474370.2 | 1 | 358 | AAN73034.1 | |
| AY598783.1 | 1 | 358 | ||
| BC041390.2 | 1 | 313 | AAH41390.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1003285 | Chr.3:31386933 - 33172886 on Build GRCh38 | Gain |
|
| nsv1011573 | Chr.3:31860029 - 32251734 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map