Assay Details
Target Gene Details
Entrez Gene ID: | 60493 |
Gene Name: | FAST kinase domains 5 |
Gene Aliases: |
dJ1187M17.5 |
Location: |
Chr.20:3146519-3159886 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FASTKD5 | NM_021826.4 | 2 | 574 | NP_068598.1 |
| AB058695.1 | 1 | 2940 | ||
| AK023211.1 | 2 | 553 | BAB14464.1 | |
| AK301748.1 | 2 | 523 | ||
| BC007413.2 | 2 | 574 | AAH07413.1 | |
| BC011856.1 | 2 | 538 | AAH11856.1 |
Target Gene Details
Entrez Gene ID: | 22888 |
Gene Name: | U-box domain containing 5 |
Gene Aliases: |
RNF37, UBCE7IP5, UIP5, hUIP5 |
Location: |
Chr.20:3107573-3159910 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBOX5 | NM_001267584.1 | NP_001254513.1 | ||
| NM_014948.3 | NP_055763.1 | |||
| NM_199415.2 | NP_955447.1 | |||
| AB020667.1 | BAA74883.2 | |||
| AK022444.1 | ||||
| AK222570.1 | BAD96290.1 | |||
| AK222876.1 | BAD96596.1 | |||
| BC000515.2 | AAH00515.1 | |||
| BC046122.1 | AAH46122.1 | |||
| BX475562.1 | ||||
| DA101925.1 |
Target Gene Details
Entrez Gene ID: | 100134015 |
Gene Name: | UBOX5 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.20:3106911-3150867 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBOX5-AS1 | NR_038395.1 | |||
| AK097516.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv585276 | Chr.20:3030542 - 3366307 on Build GRCh38 | Gain |
|
| nsv833897 | Chr.20:2990324 - 3175146 on Build GRCh38 | Loss |
|
| nsv833898 | Chr.20:3147423 - 3309891 on Build GRCh38 | Gain+Loss |
|
| nsv524482 | Chr.20:3138909 - 3158333 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map